siRNA Details
| Gene Symbol: Tecpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240617.2 | XP_006240679.1 | 6 | 1389 | ||
| XM_006240618.2 | XP_006240680.1 | 6 | 1093 | |||
| XM_006240620.2 | XP_006240682.1 | 5 | 805 | |||
| XM_008764970.1 | XP_008763192.1 | 7 | 1156 | |||
| XM_008764971.1 | XP_008763193.1 | 4 | 832 | |||
| XM_008764972.1 | XP_008763194.1 | 5 | 775 | |||

Tecpr2
Genomic Map