siRNA Details
| Gene Symbol: Ppp1r13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108062.1 | NP_001101532.1 | 5 | 641 | ||
| XM_006240634.2 | XP_006240696.1 | 7 | 1984 | |||
| XM_008764939.1 | XP_008763161.1 | 7 | 985 | |||
| XM_008764940.1 | XP_008763162.1 | 7 | 1984 | |||
| XM_008764941.1 | XP_008763163.1 | 6 | 696 | |||

Ppp1r13b
Genomic Map