siRNA Details
| Gene Symbol: Tmem179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126280.1 | NP_001119752.1 | 1 | 120 | ||
| GenBank mRNA | BC160847.1 | - | 1 | 120 | 8362921 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem179| Gene Symbol: Tmem179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126280.1 | NP_001119752.1 | 1 | 120 | ||
| GenBank mRNA | BC160847.1 | - | 1 | 120 | 8362921 | |