siRNA Details
| Gene Symbol: LOC102546572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241098.2 | XP_006241160.1 | 4 | 1804 | ||
| XM_006241099.2 | XP_006241161.1 | 4 | 2197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102546572| Gene Symbol: LOC102546572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241098.2 | XP_006241160.1 | 4 | 1804 | ||
| XM_006241099.2 | XP_006241161.1 | 4 | 2197 | |||