siRNA Details
| Gene Symbol: Sbno2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108068.1 | NP_001101538.1 | 10, 11 | 1255 | ||
| XM_006240908.2 | XP_006240970.1 | 9, 10 | 2537 | |||
| XM_006240909.1 | XP_006240971.1 | 7, 8 | 1998 | |||
| XM_008765094.1 | XP_008763316.1 | 9, 10 | 1120 | |||
| XM_008765095.1 | XP_008763317.1 | 6, 7 | 2683 | |||

Sbno2
Genomic Map