siRNA Details
| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108070.1 | NP_001101540.1 | 12 | 2253 | ||
| GenBank mRNA | BC169056.1 | - | 3 | 865 | 7932746 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dos| Gene Symbol: Dos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108070.1 | NP_001101540.1 | 12 | 2253 | ||
| GenBank mRNA | BC169056.1 | - | 3 | 865 | 7932746 | |