siRNA Details
Gene Symbol: Ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014082.1 | NP_001014104.1 | 11, 12 | 1588 | ||
XM_006240943.1 | XP_006241005.1 | 11, 12 | 1694 | |||
GenBank mRNA | BC083785.1 | - | 11, 12 | 1588 | 7096595 |
