siRNA Details
| Gene Symbol: Btbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765291.1 | XP_008763513.1 | 9 | 1489 | ||
| XM_008765292.1 | XP_008763514.1 | 8 | 1452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd11| Gene Symbol: Btbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765291.1 | XP_008763513.1 | 9 | 1489 | ||
| XM_008765292.1 | XP_008763514.1 | 8 | 1452 | |||