siRNA Details
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175598.2 | NP_783188.1 | 3 | 543 | ||
GenBank mRNA | AY177755.1 | - | 3 | 543 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175598.2 | NP_783188.1 | 3 | 543 | ||
GenBank mRNA | AY177755.1 | - | 3 | 543 |