siRNA Details
| Gene Symbol: RGD1309995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241205.2 | XP_006241267.1 | 21, 22 | 2259 | ||
| XM_008765293.1 | XP_008763515.1 | 14, 15 | 1664 | |||
| XM_235003.8 | XP_235003.3 | 21, 22 | 2259 | |||

RGD1309995
Genomic Map