siRNA Details
| Gene Symbol: Utp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191779.1 | NP_001178708.1 | 14, 15 | 1670 | ||
| XM_006241227.2 | XP_006241289.1 | 14, 15 | 1855 | |||
| GenBank mRNA | FQ225077.1 | - | 3, 4 | 429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utp20| Gene Symbol: Utp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191779.1 | NP_001178708.1 | 14, 15 | 1670 | ||
| XM_006241227.2 | XP_006241289.1 | 14, 15 | 1855 | |||
| GenBank mRNA | FQ225077.1 | - | 3, 4 | 429 | ||