siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191780.1 | NP_001178709.1 | 17, 18 | 2395 | ||
| XM_006241241.2 | XP_006241303.1 | 18, 19 | 2444 | |||
| XM_006241242.2 | XP_006241304.1 | 18, 19 | 2441 | |||

Scyl2
Genomic Map