siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191780.1 | NP_001178709.1 | 9 | 1391 | ||
| XM_006241241.2 | XP_006241303.1 | 9 | 1428 | |||
| XM_006241242.2 | XP_006241304.1 | 9 | 1428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scyl2| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191780.1 | NP_001178709.1 | 9 | 1391 | ||
| XM_006241241.2 | XP_006241303.1 | 9 | 1428 | |||
| XM_006241242.2 | XP_006241304.1 | 9 | 1428 | |||