siRNA Details
| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108082.2 | NP_001101552.2 | 5 | 821 | ||
| XM_006241253.2 | XP_006241315.1 | 5 | 1071 | |||
| GenBank mRNA | AB005540.1 | - | 5 | 786 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk17| Gene Symbol: Cdk17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108082.2 | NP_001101552.2 | 5 | 821 | ||
| XM_006241253.2 | XP_006241315.1 | 5 | 1071 | |||
| GenBank mRNA | AB005540.1 | - | 5 | 786 | ||