siRNA Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108088.1 | NP_001101558.1 | 2 | 48 | ||
| XM_006241286.2 | XP_006241348.1 | 1 | 411 | |||
| GenBank mRNA | BC161903.1 | - | 1 | 254 | 8367209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epyc| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108088.1 | NP_001101558.1 | 2 | 48 | ||
| XM_006241286.2 | XP_006241348.1 | 1 | 411 | |||
| GenBank mRNA | BC161903.1 | - | 1 | 254 | 8367209 | |