siRNA Details
| Gene Symbol: Zfc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765372.1 | XP_008763594.1 | 26 | 4972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfc3h1| Gene Symbol: Zfc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765372.1 | XP_008763594.1 | 26 | 4972 | ||