siRNA Details
| Gene Symbol: Mdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017459.2 | NP_001017459.2 | 12 | 1765 | ||
| XM_006241385.2 | XP_006241447.1 | 12 | 1802 | |||
| XM_006241387.2 | XP_006241449.1 | 12 | 1789 | |||
| XM_006241388.2 | XP_006241450.1 | 11 | 1772 | |||
| XM_008765400.1 | XP_008763622.1 | 14 | 2079 | |||
| GenBank mRNA | BC087101.1 | - | 12 | 1731 | 7192579 | |

Mdm1
Genomic Map