siRNA Details
| Gene Symbol: Arhgef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199395.1 | NP_955427.1 | 2 | 160 | ||
| XM_006241454.2 | XP_006241516.1 | 2 | 472 | |||
| XM_006241455.2 | XP_006241517.1 | 2 | 474 | |||
| XM_006241457.2 | XP_006241519.1 | 2 | 470 | |||
| XM_006241458.2 | XP_006241520.1 | 2 | 195 | |||
| GenBank mRNA | BC061879.1 | - | 2 | 160 | 5599277 | |

Arhgef25
Genomic Map