siRNA Details
| Gene Symbol: Kif5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212523.1 | NP_997688.1 | 16 | 1966 | ||
| GenBank mRNA | AY535015.1 | - | 16 | 1966 | ||
| FQ212220.1 | - | 2006 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif5a| Gene Symbol: Kif5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212523.1 | NP_997688.1 | 16 | 1966 | ||
| GenBank mRNA | AY535015.1 | - | 16 | 1966 | ||
| FQ212220.1 | - | 2006 | ||||