siRNA Details
| Gene Symbol: Zhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271056.1 | NP_001257985.1 | 3 | 2381 | ||
| XM_006241654.2 | XP_006241716.1 | 3 | 3016 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zhx2| Gene Symbol: Zhx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271056.1 | NP_001257985.1 | 3 | 2381 | ||
| XM_006241654.2 | XP_006241716.1 | 3 | 3016 | |||