siRNA Details
| Gene Symbol: Trmt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122976.2 | NP_001116448.1 | 1 | 122 | ||
| GenBank mRNA | BC079303.1 | - | 1 | 121 | 7134578 | |
| BC097456.1 | - | 1 | 122 | 7457119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt12| Gene Symbol: Trmt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122976.2 | NP_001116448.1 | 1 | 122 | ||
| GenBank mRNA | BC079303.1 | - | 1 | 121 | 7134578 | |
| BC097456.1 | - | 1 | 122 | 7457119 | ||