siRNA Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012116.1 | NP_001012116.1 | 15 | 2166 | ||
| XM_008765453.1 | XP_008763675.1 | 15 | 2386 | |||
| GenBank mRNA | BC085828.1 | - | 15 | 2166 | 7132502 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag1| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012116.1 | NP_001012116.1 | 15 | 2166 | ||
| XM_008765453.1 | XP_008763675.1 | 15 | 2386 | |||
| GenBank mRNA | BC085828.1 | - | 15 | 2166 | 7132502 | |