siRNA Details
Gene Symbol: Mroh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765606.1 | XP_008763828.1 | 16 | 1919 | ||
XM_008765607.1 | XP_008763829.1 | 16 | 2190 | |||
XM_008765608.1 | XP_008763830.1 | 16 | 3306 | |||
XM_008765609.1 | XP_008763831.1 | 16 | 3322 | |||
XM_008765610.1 | XP_008763832.1 | 16 | 2207 | |||
XM_008765611.1 | XP_008763833.1 | 16 | 1935 | |||
XM_008765612.1 | XP_008763834.1 | 17 | 2092 | |||
XM_008765613.1 | XP_008763835.1 | 15 | 1974 | |||
XM_008765614.1 | XP_008763836.1 | 16 | 1919 | |||
XM_008765615.1 | XP_008763837.1 | 16 | 1787 | |||
XM_008765616.1 | XP_008763838.1 | 15 | 1818 | |||
XM_008765617.1 | XP_008763839.1 | 16 | 1919 | |||
XM_008765618.1 | XP_008763840.1 | 16 | 1919 |
