siRNA Details
| Gene Symbol: Naprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207609.1 | NP_997492.1 | 5 | 722 | ||
| GenBank mRNA | AY214330.1 | - | 5 | 722 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naprt1| Gene Symbol: Naprt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207609.1 | NP_997492.1 | 5 | 722 | ||
| GenBank mRNA | AY214330.1 | - | 5 | 722 | ||