siRNA Details
| Gene Symbol: Card10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130554.1 | NP_001124026.1 | 18 | 2605 | ||
| XM_006241986.2 | XP_006242048.1 | 18 | 2731 | |||
| XM_006241987.2 | XP_006242049.1 | 18 | 2731 | |||
| XM_006241988.2 | XP_006242050.1 | 18 | 3646 | |||

Card10
Genomic Map