siRNA Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130503.1 | NP_001123975.1 | 9, 10 | 1062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adsl| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130503.1 | NP_001123975.1 | 9, 10 | 1062 | ||