siRNA Details
| Gene Symbol: Scube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134884.1 | NP_001128356.1 | 5, 6 | 693 | ||
| XM_006242092.2 | XP_006242154.1 | 5, 6 | 743 | |||
| XM_008765721.1 | XP_008763943.1 | 3, 4 | 763 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scube1| Gene Symbol: Scube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134884.1 | NP_001128356.1 | 5, 6 | 693 | ||
| XM_006242092.2 | XP_006242154.1 | 5, 6 | 743 | |||
| XM_008765721.1 | XP_008763943.1 | 3, 4 | 763 | |||