siRNA Details
| Gene Symbol: Fbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127547.1 | NP_001121019.1 | 4, 5 | 611 | ||
| XM_006242161.2 | XP_006242223.1 | 4, 5 | 591 | |||
| GenBank mRNA | BC161973.1 | - | 4, 5 | 573 | 9029287 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbln1| Gene Symbol: Fbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127547.1 | NP_001121019.1 | 4, 5 | 611 | ||
| XM_006242161.2 | XP_006242223.1 | 4, 5 | 591 | |||
| GenBank mRNA | BC161973.1 | - | 4, 5 | 573 | 9029287 | |