siRNA Details
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108103.1 | NP_001101573.1 | 2 | 961 | ||
XM_006242194.2 | XP_006242256.1 | 2 | 957 | |||
XM_006242196.2 | XP_006242258.1 | 2 | 957 | |||
XM_006242197.2 | XP_006242259.1 | 2 | 957 |
