siRNA Details
| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108105.1 | NP_001101575.1 | 5 | 886 | ||
| XM_006242201.2 | XP_006242263.1 | 6 | 748 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlc1| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108105.1 | NP_001101575.1 | 5 | 886 | ||
| XM_006242201.2 | XP_006242263.1 | 6 | 748 | |||