siRNA Details
| Gene Symbol: Selo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085485.1 | NP_001078954.1 | 3 | 932 | ||
| GenBank mRNA | BC166588.1 | - | 3 | 830 | 7929890 | |
| CK479732.1 | - | 2 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selo| Gene Symbol: Selo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085485.1 | NP_001078954.1 | 3 | 932 | ||
| GenBank mRNA | BC166588.1 | - | 3 | 830 | 7929890 | |
| CK479732.1 | - | 2 | 261 | |||