siRNA Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079939.1 | NP_001073408.1 | 14 | 2049 | ||
| GenBank mRNA | BC088333.1 | - | 4 | 462 | ||
| BC129102.1 | - | 14 | 2049 | 7099869 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmf2| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079939.1 | NP_001073408.1 | 14 | 2049 | ||
| GenBank mRNA | BC088333.1 | - | 4 | 462 | ||
| BC129102.1 | - | 14 | 2049 | 7099869 | ||