siRNA Details
| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729847.4 | XP_002729893.2 | 8 | 1858 | ||
| XM_006242271.2 | XP_006242333.1 | 8 | 2165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pus7l| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729847.4 | XP_002729893.2 | 8 | 1858 | ||
| XM_006242271.2 | XP_006242333.1 | 8 | 2165 | |||