siRNA Details
| Gene Symbol: Twf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008521.1 | NP_001008521.1 | 9 | 1042 | ||
| GenBank mRNA | BC086536.1 | - | 9 | 1042 | 7106678 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Twf1| Gene Symbol: Twf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008521.1 | NP_001008521.1 | 9 | 1042 | ||
| GenBank mRNA | BC086536.1 | - | 9 | 1042 | 7106678 | |