siRNA Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108112.1 | NP_001101582.1 | 16, 17 | 1976 | ||
XM_006257400.2 | XP_006257462.1 | 16, 17 | 1923 | |||
GenBank mRNA | BC166473.1 | - | 16, 17 | 1876 | 9090730 | |
FQ231766.1 | - | 8, 9 | 1013 | |||
FQ234782.1 | - | 8, 9 | 1013 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765882.1 | XP_008764104.1 | 14, 15 | 1552 |
