siRNA Details
| Gene Symbol: Nckap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108119.1 | NP_001101589.1 | 13 | 1301 | ||
| XM_008765753.1 | XP_008763975.1 | 13 | 1357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nckap1l| Gene Symbol: Nckap1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108119.1 | NP_001101589.1 | 13 | 1301 | ||
| XM_008765753.1 | XP_008763975.1 | 13 | 1357 | |||