siRNA Details
| Gene Symbol: Kbtbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108121.1 | NP_001101591.1 | 2 | 1506 | ||
| XM_006242474.2 | XP_006242536.1 | 3 | 1605 | |||
| XM_006242476.1 | XP_006242538.1 | 3 | 1494 | |||
| XM_006242477.2 | XP_006242539.1 | 3 | 1353 | |||
| XM_006242478.2 | XP_006242540.1 | 2 | 1360 | |||

Kbtbd3
Genomic Map