siRNA Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270548.1 | NP_001257477.1 | 3 | 1252 | ||
| NM_199397.2 | NP_955429.1 | 4 | 1462 | |||
| GenBank mRNA | AJ557015.1 | - | 4 | 1462 | ||
| GQ499839.1 | - | 3 | 916 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Panx1| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270548.1 | NP_001257477.1 | 3 | 1252 | ||
| NM_199397.2 | NP_955429.1 | 4 | 1462 | |||
| GenBank mRNA | AJ557015.1 | - | 4 | 1462 | ||
| GQ499839.1 | - | 3 | 916 | |||