siRNA Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191791.1 | NP_001178720.1 | 19 | 1780 | ||
| XM_006242719.2 | XP_006242781.1 | 19 | 1864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpy19l1| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191791.1 | NP_001178720.1 | 19 | 1780 | ||
| XM_006242719.2 | XP_006242781.1 | 19 | 1864 | |||