siRNA Details
| Gene Symbol: Fli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017381.1 | NP_001017381.1 | 5, 6 | 936 | ||
| GenBank mRNA | AY972521.1 | - | 5, 6 | 936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fli1| Gene Symbol: Fli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017381.1 | NP_001017381.1 | 5, 6 | 936 | ||
| GenBank mRNA | AY972521.1 | - | 5, 6 | 936 | ||