siRNA Details
| Gene Symbol: Olr1283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000518.1 | NP_001000518.1 | 1 | 708 | ||
| GenBank mRNA | X89704.1 | - | 1 | 325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1283| Gene Symbol: Olr1283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000518.1 | NP_001000518.1 | 1 | 708 | ||
| GenBank mRNA | X89704.1 | - | 1 | 325 | ||