siRNA Details
| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191792.1 | NP_001178721.1 | 7 | 1063 | ||
| XM_006242848.2 | XP_006242910.1 | 7 | 1388 | |||
| XM_006242850.2 | XP_006242912.1 | 7 | 1637 | |||
| XM_008766123.1 | XP_008764345.1 | 7 | 1391 | |||
| GenBank mRNA | FQ221553.1 | - | 7 | 1314 | ||
| FQ223998.1 | - | 7 | 1314 | |||

Ubash3b
Genomic Map