siRNA Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207610.1 | NP_997493.1 | 14 | 2359 | ||
| XM_006242938.2 | XP_006243000.1 | 15 | 2573 | |||
| GenBank mRNA | AY325198.1 | - | 14 | 2311 | ||
| BC061761.1 | - | 14 | 2359 | 5621078 | ||

Ube4a
Genomic Map