siRNA Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108142.1 | NP_001101612.1 | 26 | 3242 | ||
| XM_006242941.2 | XP_006243003.1 | 27 | 3785 | |||
| XM_006242942.2 | XP_006243004.1 | 26 | 3722 | |||
| XM_006242943.2 | XP_006243005.1 | 25 | 3709 | |||

Sidt2
Genomic Map