siRNA Details
| Gene Symbol: Ppp2r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025418.1 | NP_001020589.1 | 9, 10 | 1174 | ||
| XM_006243032.2 | XP_006243094.1 | 11, 12 | 6284 | |||
| XM_006243033.2 | XP_006243095.1 | 10, 11 | 5841 | |||
| XM_006243034.2 | XP_006243096.1 | 11, 12 | 6289 | |||
| XM_008766260.1 | XP_008764482.1 | 12, 13 | 4902 | |||
| XM_008766261.1 | XP_008764483.1 | 12, 13 | 5269 | |||
| GenBank mRNA | BC098007.1 | - | 9, 10 | 1174 | 7453957 | |

Ppp2r1b
Genomic Map