siRNA Details
| Gene Symbol: Ppp2r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025418.1 | NP_001020589.1 | 6 | 747 | ||
| XM_006243032.2 | XP_006243094.1 | 8 | 5857 | |||
| XM_006243033.2 | XP_006243095.1 | 7 | 5414 | |||
| XM_006243034.2 | XP_006243096.1 | 8 | 5862 | |||
| XM_008766260.1 | XP_008764482.1 | 9 | 4475 | |||
| XM_008766261.1 | XP_008764483.1 | 9 | 4842 | |||
| GenBank mRNA | BC098007.1 | - | 6 | 747 | 7453957 | |

Ppp2r1b
Genomic Map