siRNA Details
| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108147.1 | NP_001101617.1 | 6 | 624 | ||
| XM_008766264.1 | XP_008764486.1 | 4 | 1345 | |||
| XM_008766265.1 | XP_008764487.1 | 4 | 1345 | |||
| XM_008766266.1 | XP_008764488.1 | 4 | 1345 | |||
| XM_008766267.1 | XP_008764489.1 | 4 | 1345 | |||
| XM_008766268.1 | XP_008764490.1 | 6 | 513 | |||

Npat
Genomic Map