siRNA Details
| Gene Symbol: Elmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191579.1 | NP_001178508.1 | 3 | 233 | ||
| XM_006243083.2 | XP_006243145.1 | 3 | 277 | |||
| XM_008766269.1 | XP_008764491.1 | 3 | 315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elmod1| Gene Symbol: Elmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191579.1 | NP_001178508.1 | 3 | 233 | ||
| XM_006243083.2 | XP_006243145.1 | 3 | 277 | |||
| XM_008766269.1 | XP_008764491.1 | 3 | 315 | |||