siRNA Details
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181382.2 | NP_852047.1 | 7, 8 | 1090 | ||
GenBank mRNA | AY266116.1 | - | 7, 8 | 1090 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181382.2 | NP_852047.1 | 7, 8 | 1090 | ||
GenBank mRNA | AY266116.1 | - | 7, 8 | 1090 |