siRNA Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030039.1 | NP_001025210.1 | 9 | 916 | ||
| XM_006243163.1 | XP_006243225.1 | 10 | 1562 | |||
| XM_006243164.2 | XP_006243226.1 | 10 | 1245 | |||
| GenBank mRNA | BC098863.1 | - | 9 | 916 | 7380519 | |
| FQ227277.1 | - | 1 | 32 | |||
| X58631.1 | - | 10 | 951 | |||

Csk
Genomic Map